Imprimer

Anti-GSTM1/2 Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302987EA 547 EUR
ORIGTA302987
Anti-GSTM1/2 Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:1000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: GSTM1/2 (glutathione S-transferase mu 1/2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR