Anti-ACHE Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302997EA 438 EUR
ORIGTA302997
Anti-ACHE Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: ACHE (acetylcholinesterase (Yt blood group))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR