Anti-SEMA3E Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302998EA 438 EUR
ORIGTA302998
Anti-SEMA3E Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000 Western Blot: 2-4µg/ml

Type: Primary
Antigen: SEMA3E (sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3E)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR