Anti-RBP1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303002EA 438 EUR
ORIGTA303002
Anti-RBP1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: RBP1 (retinol binding protein 1, cellular)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR