Imprimer

Anti-SERPINI2 Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA303010EA 547 EUR
ORIGTA303010
Anti-SERPINI2 Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:64000; Western Blot: 03-1µg/ml; Immunochemistry: 3?g/ml

Type: Primary
Antigen: SERPINI2 (serpin peptidase inhibitor, clade I (pancpin), member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR