Anti-HNF4A Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303011EA 438 EUR InStock
ORIGTA303011
Anti-HNF4A Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: HNF4A (hepatocyte nuclear factor 4, alpha)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR