Anti-SERPINE1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303014EA 438 EUR
ORIGTA303014
Anti-SERPINE1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.1-0.3?g/ml; Immunochemistry: 4-6?g/ml

Type: Primary
Antigen: SERPINE1 (serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR