Imprimer

Anti-MC3R Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA303015EA 547 EUR
ORIGTA303015
Anti-MC3R Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:16000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: MC3R (melanocortin 3 receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR