Anti-GAD2 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303017EA 502 EUR
ORIGTA303017
Anti-GAD2 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: GAD2 (glutamate decarboxylase 2 (pancreatic islets and brain, 65kDa))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR