Anti-RNLS Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303023EA 438 EUR
ORIGTA303023
Anti-RNLS Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000; Western Blot: 0.1-0.3?g/ml; Immunochemistry: 2-4?g/ml

Type: Primary
Antigen: RNLS (renalase, FAD-dependent amine oxidase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR