Imprimer

Anti-PVALB Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA303033EA 438 EUR InStock
ORIGTA303033
Anti-PVALB Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:32000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: PVALB (parvalbumin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR