Anti-PPY Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303038EA 438 EUR
ORIGTA303038
Anti-PPY Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000 Western Blot: 3µg/ml

Type: Primary
Antigen: PPY (pancreatic polypeptide)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR