Anti-DAZL Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303047EA 438 EUR
ORIGTA303047
Anti-DAZL Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: DAZL (deleted in azoospermia-like)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR