Anti-GATA1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303049EA 438 EUR
ORIGTA303049
Anti-GATA1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000; Western Blot: 03-10µg/ml

Type: Primary
Antigen: GATA1 (GATA binding protein 1 (globin transcription factor 1))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR