Anti-LDB3 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303056EA 438 EUR
ORIGTA303056
Anti-LDB3 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:2000 Western Blot: 001-003µg/ml

Type: Primary
Antigen: LDB3 (LIM domain binding 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR