Anti-OSBPL10 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303066EA 438 EUR
ORIGTA303066
Anti-OSBPL10 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: OSBPL10 (oxysterol binding protein-like 10)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR