Imprimer

Anti-SNX9 Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA303091EA 547 EUR
ORIGTA303091
Anti-SNX9 Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:1000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: SNX9 (sorting nexin 9)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR