Anti-MAP3K7 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303111EA 500 EUR
ORIGTA303111
Anti-MAP3K7 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:1000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: MAP3K7 (mitogen-activated protein kinase kinase kinase 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR