Anti-COMT Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303130EA 502 EUR
ORIGTA303130
Anti-COMT Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000; Western Blot: 003-01µg/ml

Type: Primary
Antigen: COMT (catechol-O-methyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR