Anti-PAK1 Goat polyclonal antibody

Supplier: OriGene
ORIGTA303131EA 438 EUR
ORIGTA303131
Anti-PAK1 Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:4000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: PAK1 (p21 protein (Cdc42/Rac)-activated kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR