Anti-GBX2 Goat polyclonal antibody

Supplier: OriGene
ORIGTA303134EA 500 EUR
ORIGTA303134
Anti-GBX2 Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: GBX2 (gastrulation brain homeobox 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR