Imprimer

Anti-SV2A Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA303147EA 547 EUR
ORIGTA303147
Anti-SV2A Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:64000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: SV2A (synaptic vesicle glycoprotein 2A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR