Imprimer
Anti-HCST Goat polyclonal antibody
Fournisseur: OriGene
ORIGTA303159EA 454 EUR
ORIGTA303159
Anti-HCST Goat polyclonal antibody
Recommended Dilutions: ELISA: 1:128000 Western Blot: 02-06µg/ml
Type: Primary
Antigen: HCST (hematopoietic cell signal transducer)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human