Imprimer

Anti-HOXB13 Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA303162EA 550 EUR
ORIGTA303162
Anti-HOXB13 Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:128000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: HOXB13 (homeobox B13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR