Imprimer

Anti-HAP1 Goat polyclonal antibody

Fournisseur: OriGene
ORIGTA303170EA 547 EUR
ORIGTA303170
Anti-HAP1 Goat polyclonal antibody
Anticorps
Recommended Dilutions: ELISA: 1:32000 Western Blot: 01-03µg/ml

Type: Primary
Antigen: HAP1 (huntingtin-associated protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR