Imprimer

Anti-ARF1 Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA303176EA 547 EUR
ORIGTA303176
Anti-ARF1 Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:8000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: ARF1 (ADP-ribosylation factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR