Imprimer

Anti-APOM Goat polyclonal antibody

Fournisseur: OriGene
ORIGTA303181EA 547 EUR
ORIGTA303181
Anti-APOM Goat polyclonal antibody
Anticorps
Recommended Dilutions: ELISA: 1:8000 Western Blot: 3-6µg/ml

Type: Primary
Antigen: APOM (apolipoprotein M)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR