Imprimer

Anti-FGF23 Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA303182EA 547 EUR
ORIGTA303182
Anti-FGF23 Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:32000; Western Blot: 05-15µg/ml

Type: Primary
Antigen: FGF23 (fibroblast growth factor 23)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR