Anti-ACYP1 Goat polyclonal antibody

Supplier: OriGene
ORIGTA303184EA 438 EUR InStock
ORIGTA303184
Anti-ACYP1 Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:128000; Western Blot: 001-003µg/ml

Type: Primary
Antigen: ACYP1 (acylphosphatase 1, erythrocyte (common) type)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR