Imprimer

Anti-SLC25A47 Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA303188EA 547 EUR
ORIGTA303188
Anti-SLC25A47 Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:16000 Western Blot: 25µg/ml

Type: Primary
Antigen: SLC25A47 (solute carrier family 25, member 47)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR