Anti-FAIM Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303191EA 502 EUR
ORIGTA303191
Anti-FAIM Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 05-1µg/ml

Type: Primary
Antigen: FAIM (Fas apoptotic inhibitory molecule)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR