Imprimer

Anti-LARP1B Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA303192EA 550 EUR
ORIGTA303192
Anti-LARP1B Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:128000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: LARP1B (La ribonucleoprotein domain family, member 1B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR