Anti-NPY2R Goat polyclonal antibody

Supplier: OriGene
ORIGTA303196EA 438 EUR
ORIGTA303196
Anti-NPY2R Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: NPY2R (neuropeptide Y receptor Y2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR