Imprimer

Anti-SMN1 / SMN2 Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA303208EA 547 EUR
ORIGTA303208
Anti-SMN1 / SMN2 Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:1000; Western Blot: 2-4µg/ml

Type: Primary
Antigen: SMN1 / SMN2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR