Anti-ROBO3 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303233EA 500 EUR
ORIGTA303233
Anti-ROBO3 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 05-1µg/ml

Type: Primary
Antigen: ROBO3 (roundabout, axon guidance receptor, homolog 3 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR