Anti-ILF2 Goat polyclonal antibody

Supplier: OriGene
ORIGTA303236EA 438 EUR
ORIGTA303236
Anti-ILF2 Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:4000; Western Blot: 03-1µg/ml Immunofluorescence: 2-3µg/ml

Type: Primary
Antigen: ILF2 (interleukin enhancer binding factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR