Imprimer

Anti-GABPB2 Goat polyclonal antibody

Fournisseur: OriGene
ORIGTA303240EA 547 EUR
ORIGTA303240
Anti-GABPB2 Goat polyclonal antibody
Anticorps
Recommended Dilutions: ELISA: 1:32000; Western Blot: 05-15µg/ml

Type: Primary
Antigen: GABPB2 (GA binding protein transcription factor, beta subunit 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR