Anti-ITGAM Goat polyclonal antibody

Supplier: OriGene
ORIGTA303242EA 438 EUR
ORIGTA303242
Anti-ITGAM Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3 ?g/ml; Immunochemistry: 2-4 ?g/ml

Type: Primary
Antigen: ITGAM (integrin, alpha M (complement component 3 receptor 3 subunit))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR