Anti-TGM2 Goat polyclonal antibody

Supplier: OriGene
ORIGTA303243EA 438 EUR InStock
ORIGTA303243
Anti-TGM2 Goat polyclonal antibody
Antibodies
Recommended Dilutions: Western Blot: 0.03-0.1?g/ml

Type: Primary
Antigen: TGM2 (transglutaminase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR