Anti-CACNA2D1 Goat polyclonal antibody

Supplier: OriGene
ORIGTA303244EA 438 EUR
ORIGTA303244
Anti-CACNA2D1 Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000; Western Blot: 003-01µg/ml

Type: Primary
Antigen: CACNA2D1 (calcium channel, voltage-dependent, alpha 2/delta subunit 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR