Anti-APOA5 Goat polyclonal antibody

Supplier: OriGene
ORIGTA303257EA 438 EUR
ORIGTA303257
Anti-APOA5 Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: APOA5 (apolipoprotein A-V)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR