Imprimer

Anti-GOT1 Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA303294EA 547 EUR
ORIGTA303294
Anti-GOT1 Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:16000; Western Blot: 05-15µg/ml

Type: Primary
Antigen: GOT1 (glutamic-oxaloacetic transaminase 1, soluble)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR