Anti-COL4A3BP Goat polyclonal antibody

Supplier: OriGene
ORIGTA303299EA 438 EUR
ORIGTA303299
Anti-COL4A3BP Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 05-15µg/ml

Type: Primary
Antigen: COL4A3BP (collagen, type IV, alpha 3 (Goodpasture antigen) binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR