Imprimer

Anti-GOT2 Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA303304EA 547 EUR
ORIGTA303304
Anti-GOT2 Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:4000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: GOT2 (glutamic-oxaloacetic transaminase 2, mitochondrial)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR