Imprimer

Anti-FZD7 Goat polyclonal antibody

Fournisseur: OriGene
ORIGTA303332EA 550 EUR
ORIGTA303332
Anti-FZD7 Goat polyclonal antibody
Anticorps
Recommended Dilutions: ELISA: 1:64000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: FZD7 (frizzled class receptor 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR