Anti-HCK Rabbit monoclonal antibody

ORIGTA303532EA 446 EUR
ORIGTA303532
Anti-HCK Rabbit monoclonal antibody
Recommended Dilutions: Western Blot: 1:20000; Immunochemistry: 1:100; Flow Cytometry: 1:60

Type: Primary
Antigen: HCK (hemopoietic cell kinase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR