Imprimer
Anti-PIK3CD Rabbit Monoclonal Antibody [clone: Y387]
Fournisseur: OriGene
ORIGTA303639EA 810 EUR
ORIGTA303639
Anti-PIK3CD Rabbit Monoclonal Antibody [clone: Y387]
Anticorps
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:250-500; Immunoprecipitation: 1:50
Type: Primary
Antigen: PIK3CD (phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit delta)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human