Anti-RAF1 Rabbit Monoclonal Antibody [clone: Y198]

Supplier: OriGene
ORIGTA303679EA 675 EUR
ORIGTA303679
Anti-RAF1 Rabbit Monoclonal Antibody [clone: Y198]
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50-100;; Immunoprecipitation: 1:100

Type: Primary
Antigen: RAF1 (v-raf-1 murine leukemia viral oncogene homolog 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR