Anti-GRM7 Goat Antibody

Supplier: OriGene
ORIGTA305625EA 500 EUR
ORIGTA305625
Anti-GRM7 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 05-2?g/ml

Type: Primary
Antigen: GRM7 (glutamate receptor, metabotropic 7)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR