Anti-APOBEC1 Goat antibody

Supplier: OriGene
ORIGTA305633EA 500 EUR
ORIGTA305633
Anti-APOBEC1 Goat antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: APOBEC1 (apolipoprotein B mRNA editing enzyme, catalytic polypeptide 1)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR